MAN238930249 Manchester Organics

4-Aminobenzo[b]thiophene

  • MDL number MFCD01860288
  • Molecular Weight : 149.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S19695

Safety Information

MSDS https://www.manchesterorganics.com/S19695
Availability details for
Quantity
  • Availability
  • Add to Cart