MAN238930293 Manchester Organics

5-Bromo-3-morpholin-4-yl-pyrazin-2-ylamine

  • MDL number MFCD08056165
  • Molecular Weight : 259.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K20257

Safety Information

MSDS https://www.manchesterorganics.com/K20257
Availability details for
Quantity
  • Availability
  • Add to Cart