MAN238930323 Manchester Organics

3-(Tetrahydrofuran-2-yl)propionic acid

  • MDL number MFCD03412485
  • Molecular Weight : 144.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P20355

Safety Information

MSDS https://www.manchesterorganics.com/P20355
Availability details for
Quantity
  • Availability
  • Add to Cart