MAN238930327 Manchester Organics

(S)-3-Benzyloxypyrrolidine hydrochloride

  • MDL number MFCD07784038
  • Molecular Weight : 213.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S20382

Safety Information

MSDS https://www.manchesterorganics.com/S20382
Availability details for
Quantity
  • Availability
  • Add to Cart