MAN238930350 Manchester Organics

4-Cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester

  • MDL number MFCD00173757
  • Molecular Weight : 213.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L11005
Availability details for
Quantity
  • Availability
  • Add to Cart