MAN238930352 Manchester Organics

2,3,7-Trimethylindole

  • MDL number MFCD00044721
  • Molecular Weight : 159.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J17629
Availability details for
Quantity
  • Availability
  • Add to Cart