MAN238930353 Manchester Organics

2,3,5-Trimethylthiophene-4-carboxaldehyde

  • MDL number MFCD07368975
  • Molecular Weight : 154.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L17725

Safety Information

MSDS https://www.manchesterorganics.com/L17725
Availability details for
Quantity
  • Availability
  • Add to Cart