MAN238930370 Manchester Organics

4-Bromo-2-fluoro-6-methoxybenzeneboronic acid

  • MDL number MFCD09475882
  • Molecular Weight : 248.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C17671

Safety Information

MSDS https://www.manchesterorganics.com/C17671
Availability details for
Quantity
  • Availability
  • Add to Cart