MAN238930376 Manchester Organics

Benzothiazole-2-carboxylic acid

  • MDL number MFCD02682025
  • Molecular Weight : 179.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S17826

Safety Information

MSDS https://www.manchesterorganics.com/S17826
Availability details for
Quantity
  • Availability
  • Add to Cart