MAN238930396 Manchester Organics

Methyl 5-hydroxypyrazine-2-carboxylate

  • MDL number MFCD08273469
  • Cas Number : 13924-95-3
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X20197
Availability details for
Quantity
  • Availability
  • Add to Cart