MAN238930398 Manchester Organics

4-(Benzyloxy)-2-chloropyrimidine

  • MDL number MFCD06798233
  • Cas Number : 108381-28-8
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D20203
Availability details for
Quantity
  • Availability
  • Add to Cart