MAN238930405 Manchester Organics

2-Bromo-N-(1-phenylethyl)acetamide

  • MDL number MFCD05624926
  • Molecular Weight : 242.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X20646

Safety Information

MSDS https://www.manchesterorganics.com/X20646
Availability details for
Quantity
  • Availability
  • Add to Cart