MAN238930408 Manchester Organics

Malonohydrazide

  • MDL number MFCD00041268
  • Cas Number : 3815-86-9
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name malonohydrazide
Inchi Code 1S/C3H8N4O2/c4-6-2(8)1-3(9)7-5/h1,4-5H2,(H,6,8)(H,7,9)
Inchi Key PSIKPHJLTVSQFO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E20229
Availability details for
Quantity
  • Availability
  • Add to Cart