MAN238930419 Manchester Organics

2-Chloro-2,3,3,3-tetrafluoropropanoic acid

  • MDL number MFCD00465559
  • Molecular Weight : 180.48545

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T20264

Safety Information

MSDS https://www.manchesterorganics.com/T20264
Availability details for
Quantity
  • Availability
  • Add to Cart