MAN238930426 Manchester Organics

2-Benzoylcyclohexanecarboxylic acid

  • MDL number MFCD00800519
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/P20284
Availability details for
Quantity
  • Availability
  • Add to Cart