MAN238930429 Manchester Organics

[2-(2-Fluorophenyl)-2-pyrrolidin-1-ylethyl]amine

  • MDL number MFCD07186411
  • Molecular Weight : 208.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X20717

Safety Information

MSDS https://www.manchesterorganics.com/X20717
Availability details for
Quantity
  • Availability
  • Add to Cart