MAN238930432 Manchester Organics

2-(4-Hydroxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03421925
  • Molecular Weight : 279.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E20371

Safety Information

MSDS https://www.manchesterorganics.com/E20371
Availability details for
Quantity
  • Availability
  • Add to Cart