MAN238930442 Manchester Organics

4-{[2-(Methylthio)phenyl]amino}-4-oxobutanoic acid

  • MDL number MFCD06065734
  • Molecular Weight : 239.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H20397

Safety Information

MSDS https://www.manchesterorganics.com/H20397
Availability details for
Quantity
  • Availability
  • Add to Cart