MAN238930453 Manchester Organics

2,4-Dimethylquinoline-7-carboxylic acid

  • MDL number MFCD09261473
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D20416

Safety Information

MSDS https://www.manchesterorganics.com/D20416
Availability details for
Quantity
  • Availability
  • Add to Cart