MAN238930456 Manchester Organics

(2E)-3-(2-Furyl)-1-phenylprop-2-en-1-one

  • MDL number MFCD00136235
  • Molecular Weight : 198.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G20419

Safety Information

MSDS https://www.manchesterorganics.com/G20419
Availability details for
Quantity
  • Availability
  • Add to Cart