MAN238930463 Manchester Organics

Pyridine-2,5-dicarboxamide

  • MDL number MFCD00457415
  • Cas Number : 4663-96-1
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-pyridinedicarboxamide
Inchi Code 1S/C7H7N3O2/c8-6(11)4-1-2-5(7(9)12)10-3-4/h1-3H,(H2,8,11)(H2,9,12)
Inchi Key LEIAZUKOBVWTEH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S20357
Availability details for
Quantity
  • Availability
  • Add to Cart