MAN238930466 Manchester Organics

4-Amino-3-chloro-2,5,6-trifluoropyridine

  • MDL number MFCD00010856
  • Molecular Weight : 182.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K19524

Safety Information

MSDS https://www.manchesterorganics.com/K19524
Availability details for
Quantity
  • Availability
  • Add to Cart