MAN238930475 Manchester Organics

2-(Benzyloxy)-5-bromopyridine

  • MDL number MFCD09037462
  • Cas Number : 83664-33-9
  • Molecular Weight : 264.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C20202
Availability details for
Quantity
  • Availability
  • Add to Cart