MAN238930477 Manchester Organics

2,3-Dichloropyridine-4-boronic acid

  • MDL number MFCD04038756
  • Molecular Weight : 191.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H20278

Safety Information

MSDS https://www.manchesterorganics.com/H20278
Availability details for
Quantity
  • Availability
  • Add to Cart