MAN238930482 Manchester Organics

4-Cyanoquinoline

  • MDL number MFCD00972111
  • Molecular Weight : 154.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B20153
Availability details for
Quantity
  • Availability
  • Add to Cart