MAN238930486 Manchester Organics

4-Amino-N-(2-methylphenyl)benzamide

  • MDL number MFCD00443937
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W20599

Safety Information

MSDS https://www.manchesterorganics.com/W20599
Availability details for
Quantity
  • Availability
  • Add to Cart