MAN238930489 Manchester Organics

2-Bromo-N-(3-chlorophenyl)acetamide

  • MDL number MFCD00719259
  • Molecular Weight : 248.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B20272

Safety Information

MSDS https://www.manchesterorganics.com/B20272
Availability details for
Quantity
  • Availability
  • Add to Cart