MAN238930491 Manchester Organics

2-(2-Hydroxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD00411806
  • Molecular Weight : 283.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C20273

Safety Information

MSDS https://www.manchesterorganics.com/C20273
Availability details for
Quantity
  • Availability
  • Add to Cart