MAN238930535 Manchester Organics

5-Chloro-2,1,3-benzothiadiazole

  • MDL number MFCD00174269
  • Molecular Weight : 170.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M20401

Safety Information

MSDS https://www.manchesterorganics.com/M20401
Availability details for
Quantity
  • Availability
  • Add to Cart