MAN238930542 Manchester Organics

5-(Hydroxymethyl)quinolin-8-ol

  • MDL number MFCD03614757
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z20767

Safety Information

MSDS https://www.manchesterorganics.com/Z20767
Availability details for
Quantity
  • Availability
  • Add to Cart