MAN238930550 Manchester Organics

2,8-Dimethylquinoline-4-carboxylic acid

  • MDL number MFCD00487554
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D20370

Safety Information

MSDS https://www.manchesterorganics.com/D20370
Availability details for
Quantity
  • Availability
  • Add to Cart