MAN238930552 Manchester Organics

6-Chloro-2-(3-hydroxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD03421935
  • Molecular Weight : 299.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G20373

Safety Information

MSDS https://www.manchesterorganics.com/G20373
Availability details for
Quantity
  • Availability
  • Add to Cart