MAN238930557 Manchester Organics

2-Chloro-N-isopropylpropanamide

  • MDL number MFCD06384988
  • Molecular Weight : 149.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S20499

Safety Information

MSDS https://www.manchesterorganics.com/S20499
Availability details for
Quantity
  • Availability
  • Add to Cart