MAN238930633 Manchester Organics

11,12-Dihyrdoindolo[2,3-a]carbazole

  • MDL number MFCD07371389
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y20885

Safety Information

MSDS https://www.manchesterorganics.com/Y20885
Availability details for
Quantity
  • Availability
  • Add to Cart