MAN238930645 Manchester Organics

2-Phenyl-imidazo[1,2-a]pyrimidin-3-ylamine

  • MDL number MFCD07021267
  • Cas Number : 89185-48-8
  • Molecular Weight : 210.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-phenylimidazo[1,2-a]pyrimidin-3-amine
Inchi Code 1S/C12H10N4/c13-11-10(9-5-2-1-3-6-9)15-12-14-7-4-8-16(11)12/h1-8H,13H2
Inchi Key FOJZDMGNAKIZQC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z20460
Availability details for
Quantity
  • Availability
  • Add to Cart