MAN238930664 Manchester Organics

3-Bromo-2-chloroquinoline

  • MDL number MFCD08457743
  • Molecular Weight : 242.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y20956

Safety Information

MSDS https://www.manchesterorganics.com/Y20956
Availability details for
Quantity
  • Availability
  • Add to Cart