MAN238930690 Manchester Organics

5-Bromo-2-methyl-3-nitropyridine

  • MDL number MFCD09031419
  • Cas Number : 911434-05-4
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-methyl-3-nitropyridine
Inchi Code 1S/C6H5BrN2O2/c1-4-6(9(10)11)2-5(7)3-8-4/h2-3H,1H3
Inchi Key FZZLWWNOYMHSIS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z20554
Availability details for
Quantity
  • Availability
  • Add to Cart