MAN238930698 Manchester Organics

6-Methyl-2-oxo-4-thiophen-2-yl-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester

  • MDL number MFCD00124780
  • Molecular Weight : 266.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D20464

Safety Information

MSDS https://www.manchesterorganics.com/D20464
Availability details for
Quantity
  • Availability
  • Add to Cart