MAN238930743 Manchester Organics

8-Chloroquinoline

  • MDL number MFCD00047618
  • Molecular Weight : 163.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K20635

Safety Information

MSDS https://www.manchesterorganics.com/K20635
Availability details for
Quantity
  • Availability
  • Add to Cart