MAN238930755 Manchester Organics

2-Bromo-6-hydroxybenzaldehyde

  • MDL number MFCD08234722
  • Molecular Weight : 201.0174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W20599

Safety Information

MSDS https://www.manchesterorganics.com/W20599
Availability details for
Quantity
  • Availability
  • Add to Cart