MAN238930767 Manchester Organics

2-(4-Methoxy-phenyl)-pyrrolidine hydrochloride

  • MDL number MFCD06660475
  • Molecular Weight : 213.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P20639

Safety Information

MSDS https://www.manchesterorganics.com/P20639
Availability details for
Quantity
  • Availability
  • Add to Cart