MAN238930803 Manchester Organics

3-Bromo-5-methylbenzenesulfonyl chloride

  • MDL number MFCD07781544
  • Molecular Weight : 269.543

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A20626
Availability details for
Quantity
  • Availability
  • Add to Cart