MAN238930807 Manchester Organics

3-Bromo-2-chlorophenol

  • MDL number MFCD08166321
  • Molecular Weight : 207.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N20638
Availability details for
Quantity
  • Availability
  • Add to Cart