MAN238930811 Manchester Organics

Methyl 4-amino-2-chlorobenzoate

  • MDL number MFCD03840484
  • Molecular Weight : 185.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y21098

Safety Information

MSDS https://www.manchesterorganics.com/Y21098
Availability details for
Quantity
  • Availability
  • Add to Cart