MAN238930812 Manchester Organics

Ethyl 4-(trifluoromethyl)phenyl acetate

  • MDL number MFCD07779351
  • Cas Number : 721-63-1
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/A20651
Availability details for
Quantity
  • Availability
  • Add to Cart