MAN238930820 Manchester Organics

(2-Chloro-3-pyridinyl)methanol

  • MDL number MFCD01935961
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z21145

Safety Information

MSDS https://www.manchesterorganics.com/Z21145
Availability details for
Quantity
  • Availability
  • Add to Cart