MAN238930822 Manchester Organics

2-Chloro-5-amino-6-methylpyridine

  • MDL number MFCD03095220
  • Molecular Weight : 142.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S20737

Safety Information

MSDS https://www.manchesterorganics.com/S20737
Availability details for
Quantity
  • Availability
  • Add to Cart