MAN238930824 Manchester Organics

3-Fluoro-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD06659496
  • Cas Number : 869108-35-0
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C20676
Availability details for
Quantity
  • Availability
  • Add to Cart