MAN238930860 Manchester Organics

3-Cyano-5-hydroxypyridine

  • MDL number MFCD11044307
  • Cas Number : 152803-24-2
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z20719
Availability details for
Quantity
  • Availability
  • Add to Cart