MAN238930863 Manchester Organics

5-Fluoropyridine-3-carboxylic acid methyl ester

  • MDL number MFCD01692049
  • Cas Number : 455-70-9
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D20725
Availability details for
Quantity
  • Availability
  • Add to Cart